Methyl 5-Nitroimidazole-4-carboxylate

Lieferant: Apollo Scientific

OR471087-1G OR471087-5G
APOSOR471087-1GEA 357 EUR
APOSOR471087-1G APOSOR471087-5G
Methyl 5-Nitroimidazole-4-carboxylate
Methyl 5-Nitroimidazole-4-carboxylate
Formel: C₅H₅N₃O₄
Molecular Weight: 171.111 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01105422
CAS-Nummer: 20271-20-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR