Methyl 5-Nitroimidazole-4-carboxylate
Lieferant: Apollo Scientific
OR471087-1G
OR471087-5G
APOSOR471087-1GEA
357
EUR
APOSOR471087-1G
APOSOR471087-5G
Methyl 5-Nitroimidazole-4-carboxylate
Methyl 5-Nitroimidazole-4-carboxylate
Formel:
C₅H₅N₃O₄ Molecular Weight: 171.111 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01105422 CAS-Nummer: 20271-20-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...