2-(Cbz-amino)-2-methylpropanamide

Lieferant: Apollo Scientific

OR471088-1G
APOSOR471088-1GEA 601 EUR
APOSOR471088-1G
2-(Cbz-amino)-2-methylpropanamide
2-(Cbz-amino)-2-methylpropanamide
Formel: C₁₂H₁₆N₂O₃
Molecular Weight: 236,267 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25967457
CAS-Nummer: 81587-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR