6-(Bromomethyl)quinoline

Lieferant: Apollo Scientific

OR471096-1G
APOSOR471096-1GEA 397 EUR
APOSOR471096-1G
6-(Bromomethyl)quinoline
6-(Bromomethyl)quinoline
Formel: C₁₀H₈BrN
Molecular Weight: 222,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12025334
CAS-Nummer: 101279-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR