Methyl-4-aminonicotinat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-aminonicotinate

OR4711-1G OR4711-5G
APOSOR4711-1GEA 55.3 EUR
APOSOR4711-1G APOSOR4711-5G
Methyl-4-aminonicotinat 98%
Methyl-4-aminonicotinat
Formel: C₇H₈N₂O₂
Molecular Weight: 152.15 g/mol
Schmelzpunkt: 172…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00955996
CAS-Nummer: 16135-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR