Ethyl-2,6-dinitrobenzoat

Lieferant: Apollo Scientific

OR471106-5G
APOSOR471106-5GEA 601 EUR
APOSOR471106-5G
Ethyl-2,6-dinitrobenzoat
Ethyl-2,6-dinitrobenzoat
Formel: C₉H₈N₂O₆
Lagertemperatur: Raumtemperatur
MDL: MFCD06204638
CAS-Nummer: 773136-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR