2,2'-Dinitrobenzidine

Lieferant: Apollo Scientific

Synonyme: 2,2'-Dinitro-[1,1'-biphenyl]-4,4'-diamine

OR471107-5G
APOSOR471107-5GEA 1120 EUR
APOSOR471107-5G
2,2'-Dinitrobenzidine
2,2'-Dinitrobenzidine
Formel: C₁₂H₁₀N₄O₄
Molecular Weight: 274,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02752221
CAS-Nummer: 5855-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR