Methyl-1-(aminomethyl)cyclopropanecarboxylate hydrochloride

Lieferant: Apollo Scientific

Synonyme: Methyl 1-(aminomethyl)cyclopropanecarboxylate hydrochloride

OR471109-1G
APOSOR471109-1GEA 916 EUR
APOSOR471109-1G
Methyl-1-(aminomethyl)cyclopropanecarboxylate hydrochloride
Methyl-1-(aminomethyl)cyclopropanecarboxylate hydrochloride
Formel: C₆H₁₂ClNO₂
Molecular Weight: 165,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10758090
CAS-Nummer: 1170782-90-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR