Methyl 3-Amino-2-phenylpropionate hydrochloride

Lieferant: Apollo Scientific

OR471110-1G
APOSOR471110-1GEA 916 EUR
APOSOR471110-1G
Methyl 3-Amino-2-phenylpropionate hydrochloride
Methyl 3-Amino-2-phenylpropionate hydrochloride
Formel: C₁₀H₁₃NO₂·ClH
Molecular Weight: 215,677 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06740372
CAS-Nummer: 91012-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR