2-(Cbz-amino)-2-(2-pyridyl)acetic acid

Lieferant: Apollo Scientific

OR471114-5G
APOSOR471114-5GEA 1330 EUR
APOSOR471114-5G
2-(Cbz-amino)-2-(2-pyridyl)acetic acid
2-(Cbz-amino)-2-(2-pyridyl)acetic acid
Formel: C₁₅H₁₄N₂O₄
Molecular Weight: 286,283 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28144003
CAS-Nummer: 1822467-34-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR