2-Amino-2-(2-pyridyl)acetic acid dihydrochloride

Lieferant: Apollo Scientific

OR471117-5G
APOSOR471117-5GEA 1120 EUR
APOSOR471117-5G
2-Amino-2-(2-pyridyl)acetic acid dihydrochloride
2-Amino-2-(2-pyridyl)acetic acid dihydrochloride
Formel: C₇H₈N₂O₂·2ClH
Molecular Weight: 225,073 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18432492
CAS-Nummer: 1263377-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR