3-Benzylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester-3-ethyl ester
Lieferant: Apollo Scientific
Synonyme:
Ethyl 1-Boc-3-benzylpiperidine-3-carboxylate
OR471119-5G
APOSOR471119-5GEA
322
EUR
APOSOR471119-5G
3-Benzylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester-3-ethyl ester
3-Benzylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester-3-ethyl ester
Formel:
C₂₀H₂₉NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11500859 CAS-Nummer: 170842-80-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...