4-Amino-4'-hydroxybenzophenone

Lieferant: Apollo Scientific

OR471121-500MG
APOSOR471121-500MGEA 713 EUR
APOSOR471121-500MG
4-Amino-4'-hydroxybenzophenone
4-Amino-4'-hydroxybenzophenone
Formel: C₁₃H₁₁NO₂
Molecular Weight: 213,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02648435
CAS-Nummer: 14963-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR