4-Hydroxy-3'-methylbenzophenone

Lieferant: Apollo Scientific

OR471123-1G
APOSOR471123-1GEA 916 EUR
APOSOR471123-1G
4-Hydroxy-3'-methylbenzophenone
4-Hydroxy-3'-methylbenzophenone
Formel: C₁₄H₁₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10699488
CAS-Nummer: 71372-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR