7-Bromo-2-chloro-5H-pyrrolo[3,2-d]pyrimidine

Lieferant: Apollo Scientific

OR471125-1G
APOSOR471125-1GEA 804 EUR
APOSOR471125-1G
7-Bromo-2-chloro-5H-pyrrolo[3,2-d]pyrimidine
7-Bromo-2-chloro-5H-pyrrolo[3,2-d]pyrimidine
Formel: C₆H₃BrClN₃
Molecular Weight: 232,465 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22398085
CAS-Nummer: 1429882-36-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR