3-Bromo-2-chloro-5-(hydroxymethyl)pyridine

Lieferant: Apollo Scientific

OR471126-5G
APOSOR471126-5GEA 1210 EUR
APOSOR471126-5G
3-Bromo-2-chloro-5-(hydroxymethyl)pyridine
3-Bromo-2-chloro-5-(hydroxymethyl)pyridine
Formel: C₆H₅BrClNO
Molecular Weight: 222,467 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15526782
CAS-Nummer: 904745-59-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR