2-Chloro-5-nitropyrimidin-4-amine
Lieferant: Apollo Scientific
|
Gefahr
|
OR471127-5G
APOSOR471127-5GEA
150
EUR
APOSOR471127-5G
2-Chloro-5-nitropyrimidin-4-amine
2-Chloro-5-nitropyrimidin-4-amine
Formel:
C₄H₃ClN₄O₂ Schmelzpunkt: 221…226 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00127771 CAS-Nummer: 1920-66-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...