Methyl 4-Methoxypyrimidine-5-carboxylate

Lieferant: Apollo Scientific

OR471128-1G
APOSOR471128-1GEA 672 EUR
APOSOR471128-1G
Methyl 4-Methoxypyrimidine-5-carboxylate
Methyl 4-Methoxypyrimidine-5-carboxylate
Formel: C₇H₈N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD27929545
CAS-Nummer: 84332-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR