Diethyl 2-(2-Bromo-4-pyridyl)malonate

Lieferant: Apollo Scientific

OR471129-1G
APOSOR471129-1GEA 804 EUR
APOSOR471129-1G
Diethyl 2-(2-Bromo-4-pyridyl)malonate
Diethyl 2-(2-Bromo-4-pyridyl)malonate
Formel: C₁₂H₁₄BrNO₄
Molecular Weight: 316,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28154652
CAS-Nummer: 1387560-71-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR