Diethyl 2-(4-Chloro-2-nitrophenyl)malonate

Lieferant: Apollo Scientific

OR471130-1G
APOSOR471130-1GEA 896 EUR
APOSOR471130-1G
Diethyl 2-(4-Chloro-2-nitrophenyl)malonate
Diethyl 2-(4-Chloro-2-nitrophenyl)malonate
Formel: C₁₃H₁₄ClNO₆
Molecular Weight: 315,706 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06204600
CAS-Nummer: 10565-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR