N-Boc-3-(4-Pyridyl)propylamine

Lieferant: Apollo Scientific

OR471131-5G
APOSOR471131-5GEA 1080 EUR
APOSOR471131-5G
N-Boc-3-(4-Pyridyl)propylamine
N-Boc-3-(4-Pyridyl)propylamine
Formel: C₁₃H₂₀N₂O₂
Molecular Weight: 236,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07366805
CAS-Nummer: 164648-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR