N-Boc-3-(4-Pyridyl)-2-propyn-1-amine

Lieferant: Apollo Scientific

OR471132-5G
APOSOR471132-5GEA 1010 EUR
APOSOR471132-5G
N-Boc-3-(4-Pyridyl)-2-propyn-1-amine
N-Boc-3-(4-Pyridyl)-2-propyn-1-amine
Formel: C₁₃H₁₆N₂O₂
Molecular Weight: 232,278 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24447935
CAS-Nummer: 777856-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR