3-(3-Hydroxyphenyl)butanoic acid

Lieferant: Apollo Scientific

OR471133-1G
APOSOR471133-1GEA 570 EUR
APOSOR471133-1G
3-(3-Hydroxyphenyl)butanoic acid
3-(3-Hydroxyphenyl)butanoic acid
Formel: C₁₀H₁₂O₃
Molecular Weight: 180,2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20643753
CAS-Nummer: 51038-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR