3-(3-Hydroxyphenyl)pentanoic acid

Lieferant: Apollo Scientific

OR471135-1G
APOSOR471135-1GEA 519 EUR
APOSOR471135-1G
3-(3-Hydroxyphenyl)pentanoic acid
3-(3-Hydroxyphenyl)pentanoic acid
Formel: C₁₁H₁₄O₃
Molecular Weight: 194.227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28154651
CAS-Nummer: 1522045-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR