3-(3-Hydroxyphenyl)-4-methylpentanoic acid

Lieferant: Apollo Scientific

OR471136-1G
APOSOR471136-1GEA 855 EUR
APOSOR471136-1G
3-(3-Hydroxyphenyl)-4-methylpentanoic acid
3-(3-Hydroxyphenyl)-4-methylpentanoic acid
Formel: C₁₂H₁₆O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD28154649
CAS-Nummer: 1519665-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR