(1R,2R)-trans-2-(Boc-amino)cyclohexanol Hydrochlorid

Lieferant: Apollo Scientific

OR471137-5G
APOSOR471137-5GEA 56.7 EUR
APOSOR471137-5G
(1R,2R)-trans-2-(Boc-amino)cyclohexanol Hydrochlorid
(1R,2R)-trans-2-(Boc-amino)cyclohexanol Hydrochlorid
Formel: C₁₁H₂₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11656036
CAS-Nummer: 155975-19-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR