Methyl 2-(3-Oxocyclohexyl)acetate
Lieferant: Apollo Scientific
|
Achtung
|
OR471138-1G
APOSOR471138-1GEA
529
EUR
APOSOR471138-1G
Methyl 2-(3-Oxocyclohexyl)acetate
Methyl 2-(3-Oxocyclohexyl)acetate
Formel:
C₉H₁₄O₃ Molecular Weight: 170,206 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00154577 CAS-Nummer: 2808-12-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...