2-Methyl-3-(2-nitrophenyl)propanoic acid
Lieferant: Apollo Scientific
OR471140-1G
APOSOR471140-1GEA
429
EUR
APOSOR471140-1G
2-Methyl-3-(2-nitrophenyl)propanoic acid
2-Methyl-3-(2-nitrophenyl)propanoic acid
Formel:
C₁₀H₁₁NO₄ Molecular Weight: 209,199 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12794488 CAS-Nummer: 60031-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...