tert-Butyl 2-(2-Pyrrolyl)acetate
Lieferant: Apollo Scientific
OR471142-1G
APOSOR471142-1GEA
397
EUR
APOSOR471142-1G
tert-Butyl 2-(2-Pyrrolyl)acetate
tert-Butyl 2-(2-Pyrrolyl)acetate
Formel:
C₁₀H₁₅NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD20662108 CAS-Nummer: 157071-50-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...