5-Methoxy-1-methyl-1,2,3,6-tetrahydropyridine
Lieferant: Apollo Scientific
OR471143-1G
APOSOR471143-1GEA
388
EUR
APOSOR471143-1G
5-Methoxy-1-methyl-1,2,3,6-tetrahydropyridine
5-Methoxy-1-methyl-1,2,3,6-tetrahydropyridine
Formel:
C₇H₁₃NO Molecular Weight: 127,184 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13705327 CAS-Nummer: 98435-42-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...