Ethyl 2,6-Dichloropyrimidine-4-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR471145-1G
APOSOR471145-1GEA
672
EUR
APOSOR471145-1G
Ethyl 2,6-Dichloropyrimidine-4-carboxylate
Ethyl 2,6-Dichloropyrimidine-4-carboxylate
Formel:
C₇H₆Cl₂N₂O₂ Molecular Weight: 221,041 g/mol Schmelzpunkt: 32 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11044601 CAS-Nummer: 18127-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...