5-(Hydroxymethyl)thiophene-3-boronic acid

Lieferant: Apollo Scientific

Synonyme: (5-(Hydroxymethyl)thiophen-3-yl)boronic acid

OR471147-1G
APOSOR471147-1GEA 143 EUR
APOSOR471147-1G
5-(Hydroxymethyl)thiophene-3-boronic acid
5-(Hydroxymethyl)thiophene-3-boronic acid
Formel: C₅H₇BO₃S
Molecular Weight: 157,983 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27665427
CAS-Nummer: 1268683-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR