Ethyl (E)-3-Morpholinoacrylate

Lieferant: Apollo Scientific

OR471150-1G
APOSOR471150-1GEA 499 EUR
APOSOR471150-1G
Ethyl (E)-3-Morpholinoacrylate
Ethyl (E)-3-Morpholinoacrylate
Formel: C₉H₁₅NO₃
Molecular Weight: 185.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28252204
CAS-Nummer: 81239-01-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR