(2-Amino-3-pyridyl)(morpholino)methanone
Lieferant: Apollo Scientific
OR471151-1G
APOSOR471151-1GEA
448
EUR
APOSOR471151-1G
(2-Amino-3-pyridyl)(morpholino)methanone
(2-Amino-3-pyridyl)(morpholino)methanone
Formel:
C₁₀H₁₃N₃O₂ Molecular Weight: 207,229 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16678947 CAS-Nummer: 1248775-87-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...