N-Phenethylbiguanide

Lieferant: Apollo Scientific

OR471153-5G
APOSOR471153-5GEA 377 EUR
APOSOR471153-5G
N-Phenethylbiguanide
N-Phenethylbiguanide
Formel: C₁₀H₁₅N₅
Molecular Weight: 205,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00242966
CAS-Nummer: 114-86-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR