3-Ethylcyclohexanol

Lieferant: Apollo Scientific

OR471154-5G
APOSOR471154-5GEA 713 EUR
APOSOR471154-5G
3-Ethylcyclohexanol
3-Ethylcyclohexanol
Formel: C₈H₁₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD18917264
CAS-Nummer: 4534-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR