2-Isopropoxypyridine-3-carboxaldehyde

Lieferant: Apollo Scientific

Synonyme: 2-Isopropoxynicotinaldehyde

OR471157-5G
APOSOR471157-5GEA 235 EUR
APOSOR471157-5G
2-Isopropoxypyridine-3-carboxaldehyde
2-Isopropoxypyridine-3-carboxaldehyde
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06804560
CAS-Nummer: 885278-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR