Methyl 2-Benzamidoacetate
Lieferant: Apollo Scientific
OR471160-1G
APOSOR471160-1GEA
478
EUR
APOSOR471160-1G
Methyl 2-Benzamidoacetate
Methyl 2-Benzamidoacetate
Formel:
C₁₀H₁₁NO₃ Schmelzpunkt: 132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00025882 CAS-Nummer: 1205-08-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...