3-Bromo-1-(methylsulphonyl)-7-azaindole

Lieferant: Apollo Scientific

OR471161-1G
APOSOR471161-1GEA 713 EUR
APOSOR471161-1G
3-Bromo-1-(methylsulphonyl)-7-azaindole
3-Bromo-1-(methylsulphonyl)-7-azaindole
Formel: C₈H₇BrN₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD28348656
CAS-Nummer: 1936025-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR