3-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid

Lieferant: Apollo Scientific

OR471163-5G
APOSOR471163-5GEA 666 EUR
APOSOR471163-5G
3-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid
3-[(tert-Butoxycarbonyl)amino]pyridine-2-carboxylic acid
Formel: C₁₁H₁₄N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD02181061
CAS-Nummer: 569687-82-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR