(3-Nitropyridin-2-yl)methanol
Lieferant: Apollo Scientific
Synonyme:
2-(Hydroxymethyl)-3-nitropyridine
OR471164-1G
APOSOR471164-1GEA
601
EUR
APOSOR471164-1G
(3-Nitropyridin-2-yl)methanol
(3-Nitropyridin-2-yl)methanol
Formel:
C₆H₆N₂O₃ Molecular Weight: 154,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18802584 CAS-Nummer: 36625-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...