1,2-Di-p-tolylethylenediamine

Lieferant: Apollo Scientific

OR471165-1G
APOSOR471165-1GEA 306 EUR
APOSOR471165-1G
1,2-Di-p-tolylethylenediamine
1,2-Di-p-tolylethylenediamine
Formel: C₁₆H₂₀N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06658956
CAS-Nummer: 124842-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR