N'-Benzoyl-N,N-diethylthiourea

Lieferant: Apollo Scientific

OR471167-5G
APOSOR471167-5GEA 1010 EUR
APOSOR471167-5G
N'-Benzoyl-N,N-diethylthiourea
N'-Benzoyl-N,N-diethylthiourea
Formel: C₁₂H₁₆N₂OS
Molecular Weight: 236,333 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00210013
CAS-Nummer: 58328-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR