Methyl 5-amino-6-methoxynicotinate
Lieferant: Apollo Scientific
OR471168-1G
APOSOR471168-1GEA
143
EUR
APOSOR471168-1G
Methyl 5-amino-6-methoxynicotinate
Methyl 5-amino-6-methoxynicotinate
Formel:
C₈H₁₀N₂O₃ Molecular Weight: 182,177 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20697606 CAS-Nummer: 59237-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...