N-(3,5-Dimethoxybenzyl)cyclopropanamine

Lieferant: Apollo Scientific

OR471171-5G
APOSOR471171-5GEA 601 EUR
APOSOR471171-5G
N-(3,5-Dimethoxybenzyl)cyclopropanamine
N-(3,5-Dimethoxybenzyl)cyclopropanamine
Formel: C₁₂H₁₇NO₂
Molecular Weight: 207,269 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11131159
CAS-Nummer: 625435-20-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR