7-Methyl-1,4-dioxaspiro[4.5]decan-8-one

Lieferant: Apollo Scientific

OR471172-5G
APOSOR471172-5GEA 438 EUR
APOSOR471172-5G
7-Methyl-1,4-dioxaspiro[4.5]decan-8-one
7-Methyl-1,4-dioxaspiro[4.5]decan-8-one
Formel: C₉H₁₄O₃
Molecular Weight: 170,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13189957
CAS-Nummer: 702-69-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR