Methyl 1-Amino-4-ethylcyclohexanecarboxylate

Lieferant: Apollo Scientific

OR471173-1G
APOSOR471173-1GEA 916 EUR
APOSOR471173-1G
Methyl 1-Amino-4-ethylcyclohexanecarboxylate
Methyl 1-Amino-4-ethylcyclohexanecarboxylate
Formel: C₁₀H₁₉NO₂
Molecular Weight: 185,263 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11212028
CAS-Nummer: 1094555-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR