4-Benzoyl-5-methyldihydrofuran-2(3H)-one

Lieferant: Apollo Scientific

OR471174-1G
APOSOR471174-1GEA 448 EUR
APOSOR471174-1G
4-Benzoyl-5-methyldihydrofuran-2(3H)-one
4-Benzoyl-5-methyldihydrofuran-2(3H)-one
Formel: C₁₂H₁₂O₃
Molecular Weight: 204,222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28348658
CAS-Nummer: 101563-78-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR