2,6-Bis[[(R)-2-[hydroxy(diphenyl)methyl]-1-pyrrolidinyl]methyl]-4-methylphenol

Lieferant: Apollo Scientific

OR471176-1G
APOSOR471176-1GEA 1010 EUR
APOSOR471176-1G
2,6-Bis[[(R)-2-[hydroxy(diphenyl)methyl]-1-pyrrolidinyl]methyl]-4-methylphenol
2,6-Bis[[(R)-2-[hydroxy(diphenyl)methyl]-1-pyrrolidinyl]methyl]-4-methylphenol
Formel: C₄₃H₄₆N₂O₃
Molecular Weight: 638,837 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08276885
CAS-Nummer: 877395-58-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR