Ethyl 5-Bromo-6-methylpyridine-2-carboxylate
Lieferant: Apollo Scientific
Synonyme:
5-Bromo-6-methylpyridine-2-carboxylic acid ethyl ester
OR471177-1G
APOSOR471177-1GEA
855
EUR
APOSOR471177-1G
Ethyl 5-Bromo-6-methylpyridine-2-carboxylate
Ethyl 5-Bromo-6-methylpyridine-2-carboxylate
Formel:
C₉H₁₀BrNO₂ Molecular Weight: 244.085 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18382715 CAS-Nummer: 1122090-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...